


Trimethylsulphonium hydroxide 0.2 M in methanol alkylation reagent, for GC derivatisation
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:17287-03-5
:
Formula:C₃H₁₀OS
MDL Number:MFCD00216756
:94.17 g/mol
UN:1992
ADR:3,II
:GHS02
:GHS06
:GHS08
Frequently Bought Together