Order Entry
Austria
ContactUsLinkComponent
 
Built-in syringe guide
  HAMI86207
undefined
Built-in syringe guide
  HAMI86207

 

  • Ext.Ø:
    0,71 mm
  • Gauge:
    22
  • Type:
    7101 KHWG
  • Type of tip:
    3
  • Volume:
    1 µl
  • Length:
    70 mm
  • Pk:
    1

 

 

The syringe guide comes with a syringe installed from the factory. This eliminates the assembly step which can lead to damage of the smaller volume syringes. This is especially critical for 7000 series syringes which have extremely small plunger wires.