Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
Protein Assays
Accessories for for Pierce™ 660 nm protein assay kit
Accessories for for Pierce™ 660 nm protein assay kit
Accessories for for Pierce™ 660 nm protein assay kit
PIER22663
:
Thermo Fisher Scientific
:
Accessories for for Pierce™ 660 nm protein assay kit
PIER22663
:
Thermo Fisher Scientific
:
Pk:
5 g
Description:
Ionic detergent compatibility reagent for Pierce™ 660 nm protein assay kit, 5×1 g