Microplate readers, Gemini™ XPS
Catalog # MLDVXPS
Supplier: Molecular Devices
Specifications
- Detektor:Photomultiplier (R-3896)
- Lichtquelle:Xenon flash lamp (1 joule/flash)
- Wellenlängenauswahl:1,0 nm increments
- Bezeichnung:Gemini™ XPS microplate reader, multi-mode,with current version of SoftMax® Pro
- Seq:01
- Modell:Gemini™ XPS
- Temperature uniformity:<1 °C at 37 °C set point
- Wellenlängengenauigkeit:±2,0 nm
- Wavelength bandwidth:9 nm
- Messdauer:Reads a 96-well plate in <15 s, 384-well plate in <45 s
- Software:SoftMax® Pro
- Wellenlängenbereich:250 - 850 nm (excitation), 360 - 850 nm (emission)
- Gewicht:16 kg
- B×T×H:580×380×220 mm
- Inkubationsbereich:Ambient +4 °C to 45 °C
- VE:1
Specifications
About this item
The Gemini™ XPS microplate spectrofluorometer provides a flexible environment to determine the optimal excitation and emission settings for most fluorescent intensity assays. The top reading Gemini™ XPS reader has dual monochromators that allow users to utilise new and novel dyes without purchasing expensive filter sets.
- Read a variety of sample formats from 6- to 384-well microplates in endpoint, kinetic, spectral scan, and well scan modes
- Eliminate the need for identifying, purchasing, and changing out filters for wavelength optimization with dual monochromators for excitation and emission wavelength selection between 250 and 850 nm
- Up to 4 wavelength pairs can be read for endpoint and kinetic measurements
- Well scanning to report a fluorescent measurement from a single point in the centre of a microplate well to multiple points across a tissue culture well
- Thorough mixing of samples in microplate wells by user-programmable Automix feature
- Automate seamlessly with the StakMax® microplate handling stacker (available separately)
The Gemini™ XPS reader uses two scanning monochromators to determine the optimal excitation and emission settings. Assays vary depending on temperature or pH conditions; the monochromators allow users to select the optimal wavelength for any assay. When methods or fluorophores change, it takes only a few software commands to adjust the reader.
Delivery: Supplied with SoftMax® Pro data acquistion and analysis software.