Order Entry
Austria
ContactUsLinkComponent
 
Endoproteinase Glu-C protease, MS grade, Pierce™
Catalog # PIER90054
CAS Number:  
undefined
Endoproteinase Glu-C protease, MS grade, Pierce™
Catalog # PIER90054
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Description:
    Glu-C protease, MS grade
  • Pk Info:
    5×10 μg

Specifications

About this item

Glu-C protease is a highly specific endoproteinase use to improve sequence coverage in mass spectrometry protein identification applications.

  • Hydrolyses proteins specifically at the carboxyl side of glutamic acids thus being complementary to tryptic digests
  • Better protein characterisation results from overlapping peptides with complementary sequence coverage and chromatographic, ionization and fragmentation properties thereby increasing sequence coverage
  • High specific activity of greater than 500 units/mg protein
  • N-terminal arginine cleavage specificity of 90% for a complex protein sample
  • Stable since it's provided in a lyophilized format

Applications include improved sequence coverage of proteins, in-solution digestion of proteins and in-gel digestion of proteins.