Order Entry
Austria
ContactUsLinkComponent
Thorium Standard Solution, 1,000 μg/ml, 2% HNO₃
Thorium Standard Solution, 1,000 μg/ml, 2% HNO₃
Catalog # PERCN9303842
Supplier:  PerkinElmer
CAS Number:  
Thorium Standard Solution, 1,000 μg/ml, 2% HNO₃
Catalog # PERCN9303842
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1,000 μg/ml
  • Matrix:
    2% HNO₃
  • Pk:
    125 mL
  • Element:
    Thorium
  • Symbol:
    Th

Specifications

About this item

Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.

Caution: These standards are in acid solution, for full details of hazards and specification, please see www.vwr.com and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.