Calibration standard, single element, scandium for AAS and ICP-OES, pure grade
Catalog # PERCN9303798
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:1
- Element:Scandium
- Symbol:Sc
Specifications
About this item
Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.