Calibration standard, single element, Lithium for AAS and ICP-OES, pure grade
Catalog # PERCN9303781
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:1
- Pk Info:1×125 ml
- Element:Lithium
- Symbol:Li
Specifications
About this item
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
- Analyzed by ICP-OES
- Analyzed by Classical Wet Assay
- 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppm level
- All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.