DNA quantitation, SpectraMax® Quant™ dsDNA assay kits
Supplier: Molecular Devices
Certificates
About this item
SpectraMax® Quant™ dsDNA Assay kits are designed for fluorescence-based dsDNA quantitation across a broad range of concentrations.
- Tailored to meet different needs
- Highly sensitive, accurate and easy to use
- Optimised for fluorescence microplate readers, such as SpectraMax® readers
- Preconfigured protocols provided in SoftMax® Pro Software for simplified data acquisition and analysis
Quantitation of DNA is a critical step in molecular biology requiring accuracy, reliability, and the use of increasingly smaller sample volumes for applications such as next-generation sequencing. Compared to spectrophotometric DNA quantification, the fluorometric method provides key advantages such as significantly increased sensitivity, high selectivity for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA, and improved contaminant tolerance (protein and carbohydrate molecules).
Frequently Bought Together