Ion selective electrode, Ammonia, solid-state, IntelliCAL™
About this item
IntelliCAL™ ISENH4181 is a digital combination ammonium ion selective electrode (ISE) with a non refillable double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Provides fast, stable, and accurate response
- Ultimate traceability in measurement history
- User alerted when re-calibration is needed
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.
Delivery information: The electrode is supplied with calibration certificate and Basic User Manual.
The analysis package contains the IntelliCAL ISENH418101 electrode, ammonium ionic strength adjustor powder pillows (100), nitrogen-ammonia 10 mg/L standard solution (500 ml), and nitrogen-ammonia 100 mg/L standard solution (500 ml).
3 Options Available Below
Product Details & Documents
IntelliCAL™ ISENH4181 is a digital combination ammonium ion selective electrode (ISE) with a non refillable double junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Provides fast, stable, and accurate response
- Ultimate traceability in measurement history
- User alerted when re-calibration is needed
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.
Delivery information: The electrode is supplied with calibration certificate and Basic User Manual.
The analysis package contains the IntelliCAL ISENH418101 electrode, ammonium ionic strength adjustor powder pillows (100), nitrogen-ammonia 10 mg/L standard solution (500 ml), and nitrogen-ammonia 100 mg/L standard solution (500 ml).
Product Family Options
Product Information
- ConcentrationTemp.TypeMaterialElectrolyteDiaphr.ConnectionØ×LAvailabilityPrice
- HACHISENH418101IntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference systemIntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system0,018 - 9000 mg/l NH4+-N0...50 °CIntelliCAL™ ISENH4181 (ammonia), solid-stateEpoxyNon refillable driTEK gelPorous PTFE annular ring1 m cable12×220 mmLog in for availability1 * 1 ST1 200,00 €
- HACHISENH418103IntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference systemIntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system0,018 - 9000 mg/l NH4+-N0...50 °CIntelliCAL™ ISENH4181 (ammonia), solid-stateEpoxyNon refillable driTEK gelPorous PTFE annular ring3 m cable12×220 mmLog in for availability1 * 1 ST1 250,00 €
- HACHISENH4181APIntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system, analysis packageIntelliCAL™ ammonia ISE electrode ISENH4181, solid-state, Ag/AgCl reference system, analysis package0,018 - 9000 mg/l NH4+-N0...50 °CIntelliCAL™ ISENH4181 (ammonia), solid-state, analysis packageEpoxyNon refillable driTEK gelPorous PTFE annular ring1 m cable12×220 mmLog in for availability1 * 1 ST1 320,00 €
Accessories
Product Information
- Catalog #ConcentrationPkAvailabilityPrice
- Nitrogen-ammonia standard solution, 100 mg/lNitrogen-ammonia standard solution, 100 mg/lHACH2406549100 mg/l as NH₃-N500 mLLog in for availability1 * 500 mL - 45,80 €
Product Information
- Catalog #ForAvailabilityPrice
- Ammonium Ionic Strength Adjustor (ISA), 100 powder pillowsAmmonium Ionic Strength Adjustor (ISA), 100 powder pillowsHACH2980699Determining very low ammonium levelsLog in for availability1 * 1 ST - 104,00 €