Order Entry
Austria
ContactUsLinkComponent
Ponceau S 0.1 % (w/v) in 5% acetic acid staining solution
Ponceau S 0.1 % (w/v) in 5% acetic acid staining solution
Catalog # 22001.
Supplier:  Biotium
CAS Number:  
undefined
Ponceau S 0.1 % (w/v) in 5% acetic acid staining solution
Catalog # 22001.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :760.56 g/mol
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892