2,6-Dichlorophenolindophenol sodium salt hydrate 90+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1266615-56-8
Formula:C₁₂H₆Cl₂NNaO₂·xH₂O
MDL Number:MFCD00150014
:290.08 g/mol (anhy)
Storage Temperature:Ambient
EINECS:210-640-4
Specification Test Results
- Appearance:Dark green to dark blue-purple Powder
- Infrared spectrum:Conforms
- Loss on drying:≤14 % (120°C)
- Titration with HClO4:≥90.0 % (On dry substance)
Specification Test Results
Frequently Bought Together