Order Entry
Austria
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Irritant
Health
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Certificates
undefined
Irritant
Health
N-Methyl-2-pyrrolidone (NMP) 98%, pure

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4690 to 1.4710 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Color scale:
    ≤100 APHA

Specification Test Results

Related Information

Frequently Bought Together