

Dichloromethane, extra dry 99.9% stabilised, AcroSeal®
Catalog # 448371000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2320
- Pk Info:4× 25 ml
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.003 % (Coulometric)
- GC:≥99.85 %
- Stabilizer:Approx. 50 ppm Amylene
- Residue after evaporation:≤0.0005 %
- Filt.over 0.2 micron PFTE:Filtered
Specification Test Results
About this item
:
:84.93 g/mol
Boiling Pt:39…40 °C (760 mmHg)
Melting Pt:–97 °C
Density:1.325 g/cm³
UN:1593
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881