Order Entry
Austria
ContactUsLinkComponent
Ammonium dihydrogen phosphate 99% for HPLC
Ammonium dihydrogen phosphate 99% for HPLC
Catalog # ACRO447382500
CAS Number:  
undefined
Ammonium dihydrogen phosphate 99% for HPLC
Catalog # ACRO447382500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Water:
    ≤1.0 %
  • Solubility:
    (10 % in water) Clear colorless
  • UV:
    (10 % in water)
  • pH:
    4.0 to 5.0 (10 % in water)
  • Titration with NaOH:
    ≥98.5 %

Specification Test Results

About this item

 :
Melting Pt:190 °C
Density:1,8 g/cm³ (20 °C)
 :115.03 g/mol
Storage Temperature:Ambient
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396