Order Entry
Austria
ContactUsLinkComponent
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
Catalog # ACRO447362500
CAS Number:  
undefined
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
Catalog # ACRO447362500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals or granules
  • Loss on drying:
    ≤1 %
  • Solubility:
    Clear colorless
  • UV:
    (10 % in water)
  • pH:
    8.5 to 9.6 (10 % in water)
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

About this item

 :
 :174.18 g/mol
Melting Pt:340 °C
Density:2,3 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383