Order Entry
Austria
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99% for biochemistry
tri-Sodium citrate dihydrate 99% for biochemistry
Certificates
undefined
tri-Sodium citrate dihydrate 99% for biochemistry

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 M in water) Clear colorless
  • UV:
    at 260 nm A: ≤0.01
  • pH:
    7.5 to 9.0
  • Titration with HClO4:
    ≥98.5 %
  • Heavy metals (as Pb):
    0.001 %

Specification Test Results

Related Information

Frequently Bought Together