Sodium chloride 99.5% for HPLC
ACRO446212500
:
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Titration Argentometric:≥99.4 %
- UV:(10 % in water)
- pH:5.0 to 9.0
: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
:1461 °C (760 mmHg)
:801 °C
:2,16 g/cm³ (20 °C)
:Ambient
:7647-14-5
:NaCl
:MFCD00003477