

EDTA tetrasodium salt dihydrate 99%
:10378-23-1
:
:C₁₀H₁₂N₂Na₄O₈·2H₂O
:MFCD00150026
:300 °C
:≥ 100 °C
:416.2 g/mol
:GHS05
:GHS07
:Ambient
- Appearance:White Crystalline powder
- Titration Complexometric:99.0 to 102.0 % (on dry substance)
- Loss on drying:6.4 % to 10.6 %
- Solubility:(16% in water) Clear solution
Frequently Bought Together