Phenylmagnesium bromide 1.6 M in CPME (cyclopentyl methyl ether), AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-58-3
:
Formula:C₆H₅BrMg
MDL Number:MFCD00000038
:181.31 g/mol
Flash Pt:–1 °C
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to orange Solution (May develop some turbidity or precipitate)
- Titration Grignard:1.50 to 1.70 M
Specification Test Results
Related Information
Frequently Bought Together