1,1,1,3,3,3-Hexafluoro-2-propanol 99% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:920-66-1
:
Formula:C₃H₂F₆O
MDL Number:MFCD00011651
Boiling Pt:59 °C (760 mmHg)
Melting Pt:–4 °C
Density:1.596 g/cm³
Flash Pt:>100 °C
UN:3265
ADR:8,I
:168.04 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Potassium (K):≤0.5 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together