Order Entry
Austria
Orders LinkContactUsLinkComponent
1-Propanol ACS

1-Propanol ACS

Irritant
Corrosive
Flammable
1-Propanol ACS
Certificates
1-Propanol ACS

2 Options Available Below

Product Details & Documents
Synonyms:  Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
  • CAS Number:71-23-8
  • Formula:H₃CCH₂CH₂OH
  • MDL Number:MFCD00002941
  • Molecular Weight:60.1 g/mol
  • Boiling Pt:97 °C (760 mmHg)
  • Melting Pt:–127 °C
  • Density:0.8 g/cm³
  • Flash Pt:15 °C
  • Storage Temperature:Ambient
  • UN:1274
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.2 %
  • Solubility:
    (in water) Passes test
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Carbonyl compounds:
    ≤0.03 % (as C2H5CHO)
  • Isopropyl alcohol:
    ≤0.05 %
  • Titrable acid:
    ≤0.0004 meq/g
  • Ethanol:
    ≤0.01 %
  • Methanol:
    ≤0.01 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 46,60 €
    46,60 €
    1 * 500 mL
       46,60 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 75,60 €
    75,60 €
    1 * 1 L
       75,60 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.