n-Hexane 97+% ACS, meets analytical specifications of Reagent Ph. Eur. for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexyl hydride, Hexanen, Hexane
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Boiling Pt:69 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Refractive index:1.3750 to 1.3760 (20°C, 589 nm)
- Water:≤0.1 %
- Density:(20/20°C) 0.659 to 0.663
- GC:≥97.0 %
- UV:at 200 nm A: ≤0.7
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0002 meq/g
- Sulfur compounds (as S):≤0.005 %
- Water soluble titr. acid:≤0.0003 meq/g
- Thiophene:Passes test
Specification Test Results
Frequently Bought Together