Order Entry
Austria
ContactUsLinkComponent
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 97%
Irritant
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 97%
Catalog # ACRO445530010
CAS Number:  
undefined
Irritant
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 97%
Catalog # ACRO445530010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to off-white Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    91 to 96 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :266.19 g/mol
Melting Pt:91…93 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1142363-56-1
Formula:C₁₅H₂₇BO₃
MDL Number:MFCD11044439