

Nitric acid 60% in water, Technical Grade
Catalog # ACRO444400050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Titration with NaOH:59 to 63 %
Specification Test Results
About this item
:
:63.01 g/mol
Boiling Pt:122 °C (760 mmHg)
Melting Pt:–42 °C
Density:1.33 g/cm³
UN:2031
ADR:8,II
:GHS05
:GHS06
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349