2-Propanol, Technical Grade
Plastic drum, 5 L
Supplier: Thermo Fisher Scientific
2-Propanol, Technical Grade
Catalog # ACRO444250050
Supplier: Thermo Fisher Scientific
CAS Number: 67-63-0
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Boiling Pt:81…83 °C (760 mmHg)
- Melting Pt:–89.5 °C
- Density:0.785 g/cm³
- Flash Pt:12 °C
- Storage Temperature:Ambient
- UN:1219
- ADR:3,II
Documents
ACRO44425
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.2 %
- GC:≥99.75 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Acidity:≤0.001 %
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Cat. No.:ACRO444250050
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



