Dextran sulphate sodium salt, M.W. ∼ 6,500 - 10,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Dextran sulphate sodium from Dextran
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow to light brown Powder
- Loss on drying:≤10 %
- Solubility:(10 % in water) Clear to slightly hazy colorless to light yellow
- Molecular weight:6500 to 10000
- Residue after ignition:35 % to 50 %
- pH:5 to 8 (1 % solution)
- Specific optical rotation:+75° to +105° (20°C, 589 nm) (6% in water)
- Specific viscosity:0.018 to 0.032 (1.0M in NaCl, 25°C)
- Sulfur (S):16 to 20 %
Specification Test Results
Frequently Bought Together