Sodium thiosulphate pentahydrate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10102-17-7
:
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186
:248.19 g/mol
Melting Pt:48.5 °C
Density:1.72 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Iodimetric:≥98.5 %
- Total trace metal impurities:≤10 ppm
Specification Test Results
Frequently Bought Together