Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Sodium nitrate, extra pure

Irritant
Oxidizing
Sodium nitrate, extra pure
Certificates
Sodium nitrate, extra pure

3 Options Available Below

Product Details & Documents
  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Boiling Pt:380 °C (760 mmHg)
  • Melting Pt:306 °C
  • Density:2,26 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1498
  • ADR:5.1,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to white Crystalline powder or crystals
  • Loss on drying:
    ≤2 % (105°C)
  • pH:
    5.5 to 8.3 (5 % in water)
  • Titration after Ion exchange:
    ≥99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 63,00 €
    63,00 €
    1 * 1 kg
       63,00 €

  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 130,00 €
    130,00 €
    1 * 5 kg
       130,00 €

  • Fibre drum
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 346,00 €
    346,00 €
    1 * 25 kg
       346,00 €