Guanidinium thiocyanate 99+% for molecular biology
Catalog # ACRO433941000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:118 to 122 °C
- Titration Argentometric:99.0 to 101.0 %
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.05 %
- Solubility:(3 M in water) Clear colorless
- UV:at 280 nm A: ≤0.4
- pH:5.0 to 7.0 (1 M in water)
- Dnase,Rnase,Protease activity:None detected
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Zinc (Zn):≤2 ppm
Specification Test Results
About this item
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
CAS Number:593-84-0
:
Formula:NH₂C(=NH)NH₂·HSCN
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:118…122 °C
Density:1,29 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS07
:GHS05
Formula:CH₅N₃·CHNS