Order Entry
Austria
ContactUsLinkComponent
 
Dextran sulphate sodium salt, M.W. ∼ 500,000
 
undefined
Dextran sulphate sodium salt, M.W. ∼ 500,000

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Off-white to light yellow Powder
  • Loss on drying:
    ≤10 %
  • Sulfated ash:
    40 % to 50 %
  • Solubility:
    (10 % in water) Clear to slightly hazy colorless to light yellow
  • Molecular weight:
    ≥500000
  • pH:
    5 to 7.5 (1 % in water)
  • Specific optical rotation:
    +75° to +105° (20°C, 589 nm) (c=6, water)
  • Specific viscosity:
    0.10 to 0.70 (1.0M in NaCl, 25°C)
  • Sulfur (S):
    17 to 20 %

 

 

Frequently Bought Together