Order Entry
Austria
ContactUsLinkComponent
Dimethyl disulphide 99%, AcroSeal®
Toxic
Environment
Flammable
Dimethyl disulphide 99%, AcroSeal®
Catalog # 430821000.
CAS Number:  
undefined
Toxic
Environment
Flammable
Dimethyl disulphide 99%, AcroSeal®
Catalog # 430821000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5243 to 1.5263 (20°C, 589 nm)
  • Water:
    ≤0.06 %
  • GC:
    ≤1 % Methylmercaptan content

Specification Test Results

About this item

 :
 :94.2 g/mol
Boiling Pt:108…110 °C (760 mmHg)
Melting Pt:–85 °C
Density:1.0625 g/cm³
Flash Pt:16 °C
UN:2381
ADR:3,II
 :GHS06
 :GHS09
 :GHS02
Storage Temperature:Ambient
CAS Number:624-92-0
Formula:C₂H₆S₂
MDL Number:MFCD00008561