Order Entry
Austria
ContactUsLinkComponent
1,1-Dimethylbiguanide hydrochloride 97%
1,1-Dimethylbiguanide hydrochloride 97%
Catalog # ACRO429720050
CAS Number:  
undefined
1,1-Dimethylbiguanide hydrochloride 97%
Catalog # ACRO429720050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    216 to 224 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :165.625 g/mol
Melting Pt:218…223 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1115-70-4
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582