

Potassium hydroxide 1N in ethanol, AcroSeal®
Catalog # 429091000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Solution (May darken on storage)
- Strength:0.950 to 1.050 N
- Melting point:179 to 183 °C
- Titration with HClO4:≥98.0 %
Specification Test Results
About this item
:
:56.11 g/mol
Boiling Pt:78 °C (760 mmHg)
Melting Pt:–117 °C
Density:0.86 g/cm³
Flash Pt:12 °C
UN:2924
ADR:3,II
:GHS02
:GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553