
N-Succinimidyl Acrylate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38862-24-7
:
Formula:C₇H₇NO₄
MDL Number:MFCD00078261
:169.14 g/mol
Melting Pt:69 °C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to yellow Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration with TBAH:≥98.5 %
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together