Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 95%
ACRO399190010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- NMR (1H-NMR):Consistent with structure
: Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
:
:178…183 °C
:253.30 g/mol
:GHS07
:Ambient
:1149-23-1
:C₁₃H₁₉NO₄
:MFCD00005951