Order Entry
Austria
Orders LinkContactUsLinkComponent
Sand 40-100 mesh for analysis

Sand 40-100 mesh for analysis

Sand 40-100 mesh for analysis
Certificates
Sand 40-100 mesh for analysis

3 Options Available Below

Product Details & Documents
Synonyms:  Silicium dioxide, Silicium (IV) oxide, Silicon dioxide
  • CAS Number:14808-60-7
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Boiling Pt:2230 °C
  • Melting Pt:1610 °C
  • Density:2,622 to 2.645 g/cm³ (20 °C)
  •  :Density: ~ 2.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Light beige to grey Crystals
  • Particle size:
    40 to 100 mesh
  • Aluminium (Al):
    ≤250 ppm (Typical)
  • Calcium (Ca):
    ≤80 ppm (Typical)
  • Chromium (Cr):
    ≤0.5 ppm (Typical)
  • Iron (Fe):
    ≤65 ppm (Typical)
  • Potassium (K):
    ≤50 ppm (Typical)
  • Titanium (Ti):
    ≤150 ppm (Typical)
  • Silicon dioxide (SiO2):
    ≥99.80 % (Typical)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 38,50 €
    38,50 €
    1 * 250 g
       38,50 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 97,90 €
    97,90 €
    1 * 1 kg
       97,90 €

  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 395,00 €
    395,00 €
    1 * 5 kg
       395,00 €