Magnesium phosphate tribasic hydrate 98+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Magnesium phosphate hydrate, trimagnesium diphosphate hydrate
CAS Number:53408-95-0
:
Formula:H₂Mg₃O₉P₂
MDL Number:MFCD00150251
Molecular Weight:280.87 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Titration excess HCl:98.0 to 102.0 % (On ignited basis)
- Loss on ignition:≤32.0 % (425°C)
Specification Test Results
Frequently Bought Together