Order Entry
Austria
ContactUsLinkComponent
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97+%
Irritant
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97+%
Certificates
undefined
Irritant
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97+%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4620 (20°C, 589 nm)
  • GC:
    ≥97.0 %
  • Specific optical rotation:
    +96° to +107° (20°C, 589 nm) (C=1, EtOH)

Specification Test Results

Related Information

Frequently Bought Together