
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80049-61-2
:
Formula:C₁₁H₉F₆N₃PRu
MDL Number:MFCD02684569
:429.24 g/mol
ADR:6.1,III
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Yellow to orange to brown Powder
- Infrared spectrum:Conforms
- Assay:≥97.5 % (ex Ru)
Specification Test Results
Frequently Bought Together