Order Entry
Austria
ContactUsLinkComponent
Adenosine-3',5'-cyclic monophosphate (cAMP) 99+%
Corrosive
Adenosine-3',5'-cyclic monophosphate (cAMP) 99+%
Certificates
undefined
Corrosive
Adenosine-3',5'-cyclic monophosphate (cAMP) 99+%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤7 % (K.F.)
  • HPLC:
    ≥99.0 %
  • Ratio of Absorbances:
    lambda max. 260 nm, pH 7
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together