Cryopreservation cane
Supplier: Thermo Fisher Scientific
Certificates
About this item
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
- Cost effective
- Durable aluminum construction and vacuum insulation
- Narrow-mouth design minimises LN2 evaporation
- Secure locking clasp
Colour: Silver
Frequently Bought Together