ESD safety ankle boots, lace-up, Safety-Grip, Renzo XW Mid
Supplier: ELTEN
Certificates
About this item
Durable ESD safety shoes made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
- Climate-regulating, breathable textile lining
- Full length fleece inlay sole (ESD Pro), black
- Soft fleece ESD insole
- Anatomically-shaped insole and ankle support for improved stability
- PU scuff cap protects the leather upper
- Puncture resistant steel midsole
- Protective steel toecap (200 J)
- Extra wide version (XW)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.
Specifications
- Catalog No:
- 114-8168
- 114-8167
- 114-6999
- 114-8166
- 114-8175
- 114-8165
- 114-8164
- 114-8174
- 114-8163
- 114-8173
- 114-8162
- 114-8172
- 114-8171
- 114-8170
- 114-8169
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001