Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Free chlorine (DPD) reagent kit
Free chlorine (DPD) reagent kit
Free chlorine (DPD) reagent kit
Catalog # EUTE94X377001
Supplier:
EUTECH
CAS Number:
Expand image
Free chlorine (DPD) reagent kit
Catalog # EUTE94X377001
Supplier:
EUTECH
CAS Number:
Specifications
Pk:
100
Description:
Free chlorine (DPD) reagent kit
Type:
Pouches
For unit:
C401
Specifications
About this item
Product Details
Product Details