ContactUsLinkComponent
Microcentrifuge tubes
Microcentrifuge tubes
Catalog # MLBP514
   Low Manufacturing Impact
Microcentrifuge tubes
Catalog # MLBP514
   Low Manufacturing Impact
Specifications
  • Type:
    with thread, without screw cap
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Colourless
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    500
  • RCF max.:
    20000×g
Specifications
About this item
PP, translucent.
  • Frosted writing area on tube and cap