ESD safety ankle boots, lace-up, L10, Sensation Up Mid
Supplier: ELTEN
Certificates
About this item
Vintage and casual style, environmental friendly manufactured ESD safety shoe made from made from hydrophobised textile material in jeans look and hydrophobised nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
- Fabric lining Bioactive
- Replaceable insole; orthopaedic insoles possible
- Full-length inlay sole ESD PRO grey
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- 114-8831
- 114-8830
- 114-8829
- 114-8828
- 114-8827
- 114-8826
- 114-8825
- 114-8824
- 114-8823
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001