Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for pHTestr 10, 20, 30, Eutech
Accessories for pHTestr 10, 20, 30, Eutech
Accessories for pHTestr 10, 20, 30, Eutech
Catalog # EUTEPHSENSOR03DJ
Supplier:
EUTECH
Expand image
Accessories for pHTestr 10, 20, 30, Eutech
Catalog # EUTEPHSENSOR03DJ
Supplier:
EUTECH
Specifications
Description:
Replacement pH sensor for waterproof pHTestr 10, 20 and 30
Pk:
1
Specifications
About this item
Product Details
Product Details