ContactUsLinkComponent
SuperNuova+™ Multi-Position Magnetic Stirrers
SuperNuova+™ Multi-Position Magnetic Stirrers
Certificates
undefined
SuperNuova+™ Multi-Position Magnetic Stirrers
Specifications
  • Catalog No:
  • BARNSP135930-33Q
  • HERAS136030-33Q
  • No. of stirring positions:
  • 4
  • 5
  • Max. stirring capacity per stirrer H₂O:
  • 4000 ml
  • 4000 ml
  • Speed range:
  • 50 - 1200 min⁻¹
  • 50 - 1200 min⁻¹
  • Plate material:
  • Ceramic
  • Ceramic
  • W×D×H:
  • 287×411×102 mm
  • 287×411×102 mm
  • Weight:
  • 9,3 kg
  • 9,3 kg
  • Electrical:
  • 220/240 V 50/60 Hz
  • 220/240 V 50/60 Hz