Storage tubes, 2D barcoded
2D barcoded tubes, with screw cap, 2,00 ml, Yes
Catalog # MATRAB-1389/500
Supplier: THERMO MATRIX TECHNOLOGIES
Storage tubes, 2D barcoded
Catalog # MATRAB-1389/500
Supplier: THERMO MATRIX TECHNOLOGIES
Springe zu:
Product Details & Documents
The tubes are ideal for sample collection and storage, including DNA, blood, cell lines and tissue samples. A permanent 2D barcode is encapsulated onto the base of each tube to ensure long term sample identification and tracking.
- Superior sample protection
- Accurate sample tracking
- Quick sample identification
Documents
Spezifikationen
- Cap included:Yes
- Capacity:2,00 ml
- Description:2D barcoded tubes, with screw cap
- Seq:02
- Cat. No.:MATRAB-1389/500
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Produktinformationen
- Catalog #AvailabilityPrice
- Empty racks, blue for 2 ml tubes



