Cell culture media, Dulbecco's Modified Eagles Medium (DMEM), Low Glucose, HyClone™
DMEM Low Glucose was the original modification of Basal Medium Eagle (BME) used to grow mouse embryonic cells. DMEM, Low Glucose is suitable for the culturing of many immortalised cell lines from humans, monkeys, hamsters, rats, mice, chicken and fish. DMEM contains four times more amino acids, vitamins and other supplementary components than Basal Medium Eagle (BME), the base media from which DMEM was derived. As DMEM contains no growth-promoting agents or proteins, supplementation with FBS is recommended for optimal performance.
- Contains 5,6 mM glucose
- Produced with WFI quality water
- 0.1 μm sterile filtered
- Manufactured using ISO 9001:2000 certified processes
- Environmentally Preferable:
Frequently Bought Together